missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SREBF1 Rabbit, anti-Human, Clone: 1P4, Proteintech
Rabbit Recombinant Monoclonal Antibody
€ 149.00 - € 439.00
Specifications
| Antigen | SREBF1 |
|---|---|
| Clone | 1P4 |
| Concentration | 800 μg/mL |
| Applications | Western Blot, ELISA |
| Form | Liquid |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18076315
|
Proteintech
82684-3-RR-100UL |
100 μL |
€ 439.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18044475
|
Proteintech
82684-3-RR-20UL |
20 μL |
€ 149.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The SREBF1 antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human SREBF1. This antibody recognizes human antigen. The SREBF1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| SREBF1 | |
| 800 μg/mL | |
| Liquid | |
| Human | |
| bHLHd1, SREBF1, SREBP 1, SREBP 1c, SREBP1 | |
| Primary |
| 1P4 | |
| Western Blot, ELISA | |
| Rabbit | |
| 6720 | |
| IgG | |
| Store at -20°C. Stable for one year after shipment. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title