missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ssDNA Antibody (TNT-3), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-29849
This item is not returnable.
View return policy
Description
ssDNA Monoclonal specifically detects ssDNA in All Species samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| ssDNA | |
| Monoclonal | |
| 0.5mg/mL | |
| Simple Western, Flow Cytometry 5-10 μg/mL, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Reacts with single stranded DNA (ssDNA). The antibody is useful for staining nucleated cells. The antibody can also be used to identify necrotic regions of tumors and damaged tissues. By immunohistochemistry, stains heterochromatic regions of cell nucleus. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| TNT-3 | |
| Unconjugated | |
| Anti-DNA, Anti-ssDNA, Clone TNT3, Clone TNT-3, DNA DNA antibody TNT3, Necrotic marker, Single Stranded DNA, ssDNA TNT-3 monoclonal | |
| Human Raji cell nuclei. | |
| 0.1 mg | |
| Primary | |
| All species | |
| IgG3 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction