missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ssDNA Antibody (TNT-3), Novus Biologicals™
Mouse Monoclonal Antibody
€ 791.00
Specifications
| Antigen | ssDNA |
|---|---|
| Clone | TNT-3 |
| Concentration | 0.5mg/mL |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
Description
ssDNA Monoclonal specifically detects ssDNA in All Species samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| ssDNA | |
| 0.5mg/mL | |
| Unconjugated | |
| RUO | |
| Anti-DNA, Anti-ssDNA, Clone TNT3, Clone TNT-3, DNA DNA antibody TNT3, Necrotic marker, Single Stranded DNA, ssDNA TNT-3 monoclonal | |
| IgG3 | |
| Affinity Purified | |
| Reacts with single stranded DNA (ssDNA). The antibody is useful for staining nucleated cells. The antibody can also be used to identify necrotic regions of tumors and damaged tissues. By immunohistochemistry, stains heterochromatic regions of cell nucleus. |
| TNT-3 | |
| Monoclonal | |
| Mouse | |
| All species | |
| Human Raji cell nuclei. | |
| Primary | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title