Learn More
SSEA4 Monoclonal Antibody (eBioMC-813-70 (MC-813-70)), eFluor™ 570, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
€ 171.00 - € 410.00
Specifications
Antigen | SSEA4 |
---|---|
Clone | eBioMC-813-70 (MC-813-70) |
Classification | Monoclonal |
Conjugate | eFluor 570 |
Form | Liquid |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15331080
|
Affymetrix eBioscience
41-8843-82 |
100 μg |
€ 410.00
100µg |
Estimated Shipment: 13-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15321080
|
Affymetrix eBioscience
41-8843-80 |
25 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Description: The eBioMC-813-70 monoclonal antibody reacts with a glycolipid carbohydrate epitope on Stage-specific embryonic antigen-4 (SSEA-4) protein. Expression is found on human teratocarcinoma stem cells (EC), human embryonic germ cells (EG), human induced pluripotent stem (iPS) cells, and human embryonic stem cells (ES) with a decrease in expression upon differentiation. Recently expression has also been found on bone marrow mesenchymal stem cells. The eBioMC-813-70 antibody has been reported to crossreact to mouse, rabbit, canine, chicken, and Old and New World monkeys. Applications Reported: This eBioMC-813-70 (MC-813-70) antibody has been reported for use in immunocytochemistry. This product has not been validated for flow cytometric analysis. Applications Tested: This eBioMC-813-70 (MC-813-70) antibody has been tested by immunocytochemistry on fixed NTERA-2 and 2102Ep cells and can be used at less than or equal to 10 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Filter Recommendation: When using this eFluor™ 570 antibody conjugate, we recommend a filter that will capture the 570 emission wavelength (for example, Excitation 545/25, 565LP, Emission 605/70). A standard Alexa Fluor™ 555 or TRITC filter is acceptable. Excitation: 555 nm; Emission: 570 nm
SSEA4 Monoclonal antibody specifically detects SSEA4 in Canine, Chicken, Human, Mouse, Non-human Primate, Rabbit samples. It is validated for Immunocytochemistry.Specifications
SSEA4 | |
Monoclonal | |
Liquid | |
RUO | |
IgG3 | |
Affinity chromatography |
eBioMC-813-70 (MC-813-70) | |
eFluor 570 | |
Mouse | |
Canine, Chicken, Human, Mouse, Non-human Primate, Rabbit | |
Primary | |
Antibody |
For Research Use Only.