missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SSH3 Mouse, anti-Human, Clone: 1A9C9, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68583-1-IG-20UL
This item is not returnable.
View return policy
Description
The SSH3 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human SSH3. This antibody recognizes human antigen. The SSH3 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| SSH3 | |
| 1A9C9 | |
| FLJ10928, FLJ20515, hSSH 3L, SSH 3, SSH 3L, SSH like protein 3, SSH3, SSH3L | |
| 20 μL | |
| 54961 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
| Western Blot, ELISA | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction