missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SSH3 Mouse, anti-Human, Clone: 1A9C9, Proteintech
Mouse Monoclonal Antibody
€ 145.00 - € 410.00
Specifications
| Antigen | SSH3 |
|---|---|
| Clone | 1A9C9 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot, ELISA |
| Form | Liquid |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18045515
|
Proteintech
68583-1-IG-20UL |
20 μL |
€ 145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18016915
|
Proteintech
68583-1-IG-150UL |
150 μL |
€ 410.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The SSH3 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human SSH3. This antibody recognizes human antigen. The SSH3 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| SSH3 | |
| 1000 μg/mL | |
| Liquid | |
| Human | |
| FLJ10928, FLJ20515, hSSH 3L, SSH 3, SSH 3L, SSH like protein 3, SSH3, SSH3L | |
| Primary |
| 1A9C9 | |
| Western Blot, ELISA | |
| Mouse | |
| 54961 | |
| IgG2b | |
| Store at -20°C. Stable for one year after shipment. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title