Learn More
SSR1 Monoclonal Antibody (OTI 5D5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA503265
This item is not returnable.
View return policy
Description
The signal sequence receptor (SSR) is a glycosylated endoplasmic reticulum (ER) membrane receptor associated with protein translocation across the ER membrane. The SSR consists of 2 subunits, a 34-kD glycoprotein encoded by this gene and a 22-kD glycoprotein. This gene generates several mRNA species as a result of complex alternative polyadenylation. This gene is unusual in that it utilizes arrays of polyA signal sequences that are exclusively non-canonical.Specifications
| SSR1 | |
| Monoclonal | |
| 0.29 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P43307 | |
| SSR1 | |
| Full length human recombit protein of human SSR1 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Western Blot | |
| OTI 5D5 | |
| Unconjugated | |
| SSR1 | |
| TRAPA | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 6745 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |