missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SSR1 Monoclonal Antibody (OTI 5D5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
€ 242.00 - € 540.87
Specifications
| Antigen | SSR1 |
|---|---|
| Clone | OTI 5D5 |
| Concentration | 0.29 mg/mL |
| Applications | Flow Cytometry, Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17588901
|
Origene Technologies
TA503265S |
30 μL |
€ 242.00
30µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17568901
|
Origene Technologies
TA503265 |
100 μL |
€ 540.87
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SSR1 Monoclonal antibody specifically detects SSR1 in Canine, Human samples. It is validated for Flow Cytometry, Western BlotSpecifications
| SSR1 | |
| 0.29 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| Primary | |
| SSR1 |
| OTI 5D5 | |
| Flow Cytometry, Western Blot | |
| Unconjugated | |
| Mouse | |
| IgG1 | |
| -20° C, Avoid Freeze/Thaw Cycles |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title