missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SSSCA1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12273-1-AP-150UL
This item is not returnable.
View return policy
Description
The calculated MW of SSSCA1 is 21.5 kDa (native form), with modification the MW is about 27 kDa.
Might play a role in mitosis. Antigenic molecule. Could be a centromere-associated protein. May induce anti-centromere antibodies.Specifications
| SSSCA1 | |
| Polyclonal | |
| Unconjugated | |
| SSSCA1 | |
| Autoantigen p27, p27, SSSCA1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10534, 56390, 689397 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O60232, P56873 | |
| Sssca1 | |
| SSSCA1 Fusion Protein Ag2916 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction