missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SSSCA1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
€ 145.00 - € 410.00
Specifications
| Antigen | SSSCA1 |
|---|---|
| Concentration | 0.27 mg/mL |
| Applications | Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Immunocytochemistry, Flow Cytometry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16819893
|
Proteintech
12273-1-AP-20UL |
20 μL |
€ 145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16809893
|
Proteintech
12273-1-AP-150UL |
150 μL |
€ 410.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The calculated MW of SSSCA1 is 21.5 kDa (native form), with modification the MW is about 27 kDa.
Might play a role in mitosis. Antigenic molecule. Could be a centromere-associated protein. May induce anti-centromere antibodies.Specifications
| SSSCA1 | |
| Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Immunocytochemistry, Flow Cytometry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| O60232, P56873 | |
| 10534, 56390, 689397 | |
| SSSCA1 Fusion Protein Ag2916 | |
| Primary | |
| -20°C | |
| SSSCA1 |
| 0.27 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Autoantigen p27, p27, SSSCA1 | |
| Sssca1 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title