Learn More
SSSCA1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 12273-1-AP-20UL
 View more versions of this product
Product Code. 16819893
  • € 145.00 / 20µL
Description
The calculated MW of SSSCA1 is 21.5 kDa (native form), with modification the MW is about 27 kDa.
Might play a role in mitosis. Antigenic molecule. Could be a centromere-associated protein. May induce anti-centromere antibodies.
Specifications
Specifications
SSSCA1
Polyclonal
Unconjugated
SSSCA1
Autoantigen p27, p27, SSSCA1
Rabbit
Antigen Affinity Chromatography
RUO
10534, 56390, 689397
-20°C
Liquid
Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Immunocytochemistry, Flow Cytometry
0.27 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O60232, P56873
Sssca1
SSSCA1 Fusion Protein Ag2916
20 μL
Primary
Human, Mouse, Rat