missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ ST7/LRP12 Antibody (7B8A3), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-27137-0.1ml
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
ST7/LRP12 Monoclonal antibody specifically detects ST7/LRP12 in Human samples. It is validated for ELISA,Western Blot
Specifikationer
| ST7/LRP12 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500 - 1:2000, ELISA 1:10000 | |
| DKFZp762O2113, Protein FAM4A1, Protein HELG, SEN4, subfamily A, member 1, suppression of tumorigenicity 7, suppression of tumorigenicity 7 (breast), suppressor of tumorigenicity 7 protein | |
| Purified recombinant fragment of human ST7/LRP12 (AA: extra 355-492) expressed in E. Coli. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| ELISA, Western Blot | |
| 7B8A3 | |
| Unconjugated | |
| PBS | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 29967 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction