missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ ST7/LRP12 Antibody (7B8A3), Novus Biologicals™
Mouse Monoclonal Antibody
€ 287.00 - € 511.00
Specifications
| Antigen | ST7/LRP12 |
|---|---|
| Clone | 7B8A3 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500 - 1:2000, ELISA 1:10000 |
| Applications | ELISA, Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30039379
|
Novus Biologicals™
NBP3-27137-0.025ml |
0.025 mL |
€ 287.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30032496
|
Novus Biologicals™
NBP3-27137-0.1ml |
0.1 mL |
€ 511.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ST7/LRP12 Monoclonal antibody specifically detects ST7/LRP12 in Human samples. It is validated for ELISA,Western BlotSpecifications
| ST7/LRP12 | |
| 1 mg/mL | |
| ELISA, Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| DKFZp762O2113, Protein FAM4A1, Protein HELG, SEN4, subfamily A, member 1, suppression of tumorigenicity 7, suppression of tumorigenicity 7 (breast), suppressor of tumorigenicity 7 protein | |
| Purified recombinant fragment of human ST7/LRP12 (AA: extra 355-492) expressed in E. Coli. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| 7B8A3 | |
| Western Blot 1:500 - 1:2000, ELISA 1:10000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS | |
| 29967 | |
| IgG1 | |
| Protein G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title