missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Stainless Steel, 1.4207, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Stainless Steel, 1.4207, Ø40x20mm
Specifications
Specifications
| Chemical Name or Material | Phosphorus/Cobalt/Nitrogen/Carbon/Tantalum/Copper/Niobium/Molybdenum/Manganese/Silicon/Tungsten/Nickel/Chromium |
| CAS | 7439-89-6 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Phosphorus 0.006 %/Cobalt 0.01 %/Nitrogen 0.01 %/Carbon 0.037 %/Tantalum 0.08 %/Copper 0.18 %/Niobium 0.26 %/Molybdenum 0.57 %/Manganese 0.58 %/Silicon 1.4 %/Tungsten 3 %/Nickel 12.3 %/Chromium 19.6 % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction