missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Stainless Steel, AISI 306, S30600, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Stainless Steel, AISI 306, S30600, Ø32x20mm
Specifications
Specifications
| Chemical Name or Material | Magnesium/Molybdenum/Carbon/Phosphorus/Copper/Cobalt/Aluminium/Vanadium/Nitrogen/Manganese/Silicon/Nickel/Chromium/Niobium |
| CAS | 7439-89-6 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Magnesium 0.0016 %/Molybdenum 0.004 %/Carbon 0.009 %/Phosphorus 0.0091 %/Copper 0.0121 %/Cobalt 0.0149 %/Aluminium 0.0204 %/Vanadium 0.0206 %/Nitrogen 0.027 %/Manganese 0.648 %/Silicon 4.23 %/Nickel 14.96 %/Chromium 17.43 %/Niobium (0.004) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction