missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Stainless Steel, AISI 347H, S34700, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Stainless Steel, AISI 347H, S34700, Ø38x19mm
Specifications
Specifications
| Chemical Name or Material | Aluminium/Titanium/Tin/Phosphorus/Carbon/Nitrogen/Vanadium/Tungsten/Cobalt/Silicon/Molybdenum/Copper/Niobium/Manganese/Nickel/Chromium/Calcium/Selenium/Zinc/Antimony/Tantalum/Zirconium/Arsenic/Lead/Boron/Oxygen |
| CAS | 7439-89-6 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Aluminium 0.003 %/Titanium 0.008 %/Tin 0.012 %/Phosphorus 0.0352 %/Carbon 0.0424 %/Nitrogen 0.052 %/Vanadium 0.057 %/Tungsten 0.06 %/Cobalt 0.301 %/Silicon 0.38 %/Molybdenum 0.416 %/Copper 0.441 %/Niobium 0.6 %/Manganese 1.395 %/Nickel 9.01 %/Chromium 17.08 %/Calcium (<0.001) %/Selenium (<0.001) %/Zinc (<0.001) %/Antimony (<0.002) %/Tantalum (<0.005) %/Zirconium (<0.005) %/Arsenic (<0.008) %/Lead (<0.03) %/Boron (0.0005) %/Oxygen (0.004) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction