missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Stainless Steel, AISI 440C, S44004, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Stainless Steel, AISI 440C, S44004, Ø40x15mm
Specifications
Specifications
| Chemical Name or Material | Niobium/Tungsten/Aluminium/Cobalt/Phosphorus/Nitrogen/Vanadium/Copper/Nickel/Manganese/Silicon/Molybdenum/Carbon/Chromium/Titanium |
| CAS | 7439-89-6 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Niobium 0.008 %/Tungsten 0.0156 %/Aluminium 0.016 %/Cobalt 0.0167 %/Phosphorus 0.0232 %/Nitrogen 0.0308 %/Vanadium 0.0484 %/Copper 0.0687 %/Nickel 0.197 %/Manganese 0.378 %/Silicon 0.44 %/Molybdenum 0.468 %/Carbon 1.012 %/Chromium 16.5 %/Titanium (0.004) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction