Learn More
B Braun™ Stainless Steel Angled Tip Watchmaker Forceps
Use B Braun™ Stainless Steel Angled Tip Watchmaker Forceps as an integral tool in a surgical toolkit.
Brand:  B Braun™ BD331R
Additional Details : Weight : 0.99000kg
 View more versions of this product
Product Code. 10094647
  • € 33.70 / Each
Estimated Shipment: 17-05-2024
to see stock.
Add to basket
Description
Description
  • Stainless steel
  • Angled tip
  • Watchmaker style
Specifications
Specifications
Angled
Stainless Steel
Forceps, Watchmaker
Forceps, Watchmaker
1 Pack
110 mm
0.15 mm