missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Stainless Steel, AISI 310, S31000, 100g, ARMI∣MBH
Click to view available options
Quantity:
100 g
Unit Size:
Each
Description
Stainless Steel, AISI 310, S31000, 100g
Specifications
Specifications
| Chemical Name or Material | Zirconium/Oxygen/Boron/Arsenic/Magnesium/Aluminium/Niobium/Tantalum/Tin/Tungsten/Phosphorus/Titanium/Carbon/Nitrogen/Cobalt/Vanadium/Copper/Molybdenum/Silicon/Manganese/Nickel/Chromium/Lead/Zinc/Antimony/Calcium/Selenium |
| CAS | 7439-89-6 |
| Quantity | 100 g |
| Form | Pieces |
| Concentration or Composition (by Analyte or Components) | Zirconium 0.002 %/Oxygen 0.003 %/Boron 0.0032 %/Arsenic 0.005 %/Magnesium 0.007 %/Aluminium 0.008 %/Niobium 0.008 %/Tantalum 0.008 %/Tin 0.008 %/Tungsten 0.017 %/Phosphorus 0.027 %/Titanium 0.029 %/Carbon 0.0454 %/Nitrogen 0.058 %/Cobalt 0.085 %/Vanadium 0.092 %/Copper 0.32 %/Molybdenum 0.58 %/Silicon 0.63 %/Manganese 1.36 %/Nickel 19.2 %/Chromium 24.9 %/Lead (0.0005) %/Zinc (0.0005) %/Antimony (0.001) %/Calcium (0.001) %/Selenium (0.001) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction