missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Stainless Steel, Custom 455, S45500, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Stainless Steel, Custom 455, S45500, Ø38x20mm
Specifications
Specifications
| Chemical Name or Material | Nitrogen/Carbon/Phosphorus/Cobalt/Molybdenum/Manganese/Silicon/Vanadium/Aluminium/Titanium/Copper/Nickel/Chromium |
| CAS | 7439-89-6 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Nitrogen 0.002 %/Carbon 0.003 %/Phosphorus 0.005 %/Cobalt 0.015 %/Molybdenum 0.015 %/Manganese 0.027 %/Silicon 0.055 %/Vanadium 0.065 %/Aluminium 0.07 %/Titanium 1.2 %/Copper 2.2 %/Nickel 8.3 %/Chromium 11.4 % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction