missing translation for 'onlineSavingsMsg'
Learn More

Stainless Steel, Custom 455, S45500, ARMI∣MBH

Product Code. 17904416
Change view
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17904416 1 Ea. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17904416 Supplier ARMI/MBH Supplier No. IARM16CX

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ARMI∣MBH produces high-quality metal alloy certified reference materials and set up standards for Spark-OES, ICP, and XRF elemental analysis. The offering includes steel, aluminum, titanium, nickel, cobalt, copper, magnesium, and more.

Stainless Steel, Custom 455, S45500, Ø38x3mm
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Zirconium/Boron/Oxygen/Arsenic/Carbon/Nitrogen/Tin/Phosphorus/Tungsten/Molybdenum/Cobalt/Manganese/Silicon/Vanadium/Aluminium/Niobium/Titanium/Copper/Nickel/Chromium
CAS 7439-89-6
Quantity 1 Ea.
Form Disc
Concentration or Composition (by Analyte or Components) Zirconium 0.001 %/Boron 0.0011 %/Oxygen 0.0014 %/Arsenic 0.003 %/Carbon 0.003 %/Nitrogen 0.003 %/Tin 0.003 %/Phosphorus 0.007 %/Tungsten 0.008 %/Molybdenum 0.009 %/Cobalt 0.017 %/Manganese 0.024 %/Silicon 0.03 %/Vanadium 0.07 %/Aluminium 0.072 %/Niobium 0.248 %/Titanium 1.16 %/Copper 2.08 %/Nickel 8.23 %/Chromium 11.34 %
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.