missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Stainless Steel, Duplex, LDX 2101, S32101, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Stainless Steel, Duplex, LDX 2101, S32101, Ø38x3mm
Specifications
Specifications
| Chemical Name or Material | Boron/Oxygen/Tin/Titanium/Niobium/Aluminium/Tungsten/Phosphorus/Carbon/Cobalt/Vanadium/Molybdenum/Nitrogen/Copper/Silicon/Nickel/Manganese/Chromium/Tantalum |
| CAS | 7439-89-6 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Boron 0.0011 %/Oxygen 0.0024 %/Tin 0.004 %/Titanium 0.005 %/Niobium 0.009 %/Aluminium 0.01 %/Tungsten 0.01 %/Phosphorus 0.018 %/Carbon 0.03 %/Cobalt 0.031 %/Vanadium 0.084 %/Molybdenum 0.097 %/Nitrogen 0.245 %/Copper 0.29 %/Silicon 0.75 %/Nickel 1.47 %/Manganese 5 %/Chromium 21.35 %/Tantalum (0.006) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction