Learn More
Stainless Steel gauze, 20 mesh woven from 0.382mm (0.015in) dia wire, Type 304, Thermo Scientific Chemicals
Filtration, electrodes
€ 33.70 - € 161.00
Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11385257
View Documents Promotion Details
Thermo Scientific Alfa Aesar
040942.FL
75 x 75 mm
€ 33.70
Each
View alternative products
Save Up to 
»
In Stock
to see stock.
11305267
View Documents Promotion Details
Thermo Scientific Alfa Aesar
040942.GT
150 x 150 mm
€ 63.30
Each
View alternative products
Save Up to 
»
In Stock
to see stock.
11395257
View Documents Promotion Details
Thermo Scientific Alfa Aesar
040942.HB
300 x 300 mm
€ 161.00
Each
View alternative products
Save Up to 
»
In Stock
to see stock.
Description
Description
Stainless Steel gauze, Type 316 is used to woven wire cloth which has extensive use in chemical processing when better corrosion resistance is required than the regular chromium-nickel types. It is also used in medical, pharmaceutical, laboratory, semiconductor, food processing, marine, waste water and water purification equipment.