missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Stainless Steel, Martensitic, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Stainless Steel, Martensitic, Ø40x20mm
Specifications
Specifications
| Chemical Name or Material | Tin/Carbon/Aluminium/Vanadium/Phosphorus/Cobalt/Copper/Molybdenum/Silicon/Niobium/Manganese/Nickel/Chromium |
| CAS | 7439-89-6 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Tin 0.001 %/Carbon 0.01 %/Aluminium 0.03 %/Vanadium 0.03 %/Phosphorus 0.06 %/Cobalt 0.15 %/Copper 0.17 %/Molybdenum 0.54 %/Silicon 0.57 %/Niobium 0.7 %/Manganese 1.42 %/Nickel 1.96 %/Chromium 18 % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction