Learn More
Tenak™ Standard Rack for Upright Freezer, 50 mm Box Height
Standard Rack fits into any upright freezer and can be used with all standard cryoboxes.
Brand:  Tenak™ TE23156
Additional Details : Weight : 4.51000kg
 View more versions of this product
Product Code. 15230674
  • € 128.00 / Each
Estimated Shipment: 10-09-2024
to see stock.
Add to basket
Description
Description
  • Measure the inside height and depth of your freezer when choosing the right rack
  • Racks can be used with all freezer brands
  • Constructed of stainless steel
Specifications
Specifications
6 x 5
Stainless Steel
683 x 139 x 331 mm
Boxes (30)
Upright Freezers
50 mm (Box)