Learn More
Ismatec™ Standard Tubing Connectors, Steel
Variously sized, standard straight tubing connectors. Ismatec™ Standard Tubing Connectors, Steel are available in different diameters and lengths.
Brand:  Ismatec™ ISM585A
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 10194542
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • Standard, non-reducing.
  • Designed for Ismatec™ pump tubing.
  • Available in different lengths and diameters.
  • Steel construction.
Specifications
Specifications
Standard Tubing Connector
25 mm
6/Pk.
Straight
Steel
For 0.30mm I.D., 0.63mm O.D. Tubing
0.30mm I.D., 0.63 O.D.