Learn More
Ismatec™ Standard Tubing Connectors, Steel
Variously sized, standard straight tubing connectors. Ismatec™ Standard Tubing Connectors, Steel are available in different diameters and lengths.
Brand: Ismatec™ ISM586A
Additional Details : Weight : 0.10000kg
Description
- Standard, non-reducing.
- Designed for Ismatec™ pump tubing.
- Available in different lengths and diameters.
- Steel construction.
Specifications
Standard Tubing Connector | |
25 mm | |
6/Pk. | |
Straight |
Steel | |
For 0.58mm I.D., 0.90mm O.D. Tubing | |
0.58mm I.D., 0.90mm O.D. |