Learn More
Stat3 (pY705) Mouse, PE, Clone: 4/P-STAT3, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 562072
Additional Details : Weight : 0.01000kg
Description
STAT (Signal transducer and activator of transcription) of Y705 in Stat3 occurs in response to growth factors and cytokines, and is essential for normal transcription activity.
The 4/P-STAT3 monoclonal antibody recognizes the phosphorylated Y705 of Stat3.
Specifications
Stat3 (pY705) | |
Monoclonal | |
PE | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Phosphorylated Human Stat3 Peptide | |
250 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
4/P-STAT3 | |
Acute-phase response factor, APRF | |
Mouse | |
Affinity Purified | |
RUO | |
Human, Mouse | |
IgG2a κ |
For Research Use Only.