missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STAT5a/b, p Tyr694 Antibody (SR2280), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21532-50ul
This item is not returnable.
View return policy
Description
STAT5a/b Monoclonal antibody specifically detects STAT5a/b in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| STAT5a/b | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, Immunohistochemistry | |
| SR2280 | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 | |
| MGF, STAT5, STAT5A signal transducer and activator of transcription 5A | |
| A synthesized peptide derived from human phospho-Stat5 [Tyr694] (Uniprot #: P42229, P51692) | |
| 50 μg | |
| Cancer, Phospho Specific, Signal Transduction, Transcription Factors and Regulators | |
| 6776 | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction