missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STAT5a/b, p Tyr694 Antibody (SR2280), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 349.00 - € 472.00
Specifications
| Antigen | STAT5a/b |
|---|---|
| Clone | SR2280 |
| Dilution | Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18661885
|
Novus Biologicals
NBP3-21532-100ul |
100 μg |
€ 472.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18604234
|
Novus Biologicals
NBP3-21532-50ul |
50 μg |
€ 349.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
STAT5a/b Monoclonal antibody specifically detects STAT5a/b in Human, Mouse samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
| STAT5a/b | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| MGF, STAT5, STAT5A signal transducer and activator of transcription 5A | |
| A synthesized peptide derived from human phospho-Stat5 [Tyr694] (Uniprot #: P42229, P51692) | |
| Primary | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. |
| SR2280 | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Cancer, Phospho Specific, Signal Transduction, Transcription Factors and Regulators | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 6776 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title