Learn More
Velp Scientifica™ Magnetic Stir Bars
Product Code. 13122172 Shop All Velp Products
Change view
Click to view available options
Dimensions (L x W x H):
6 x 20 mm
6 x 35 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W x H) unitSize
13122172 6 x 35 mm Each
13132172 6 x 20 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 13122172 Supplier Velp Scientifica™ Supplier No. A00001056
Only null left
Add to basket
Add to basket
Provides stable, general purpose stirring. Velp Scientifica™ Magnetic Stir Bars fit with a range of Velp Scientifica™ stirrer units.
  • Available in two different sizes
  • Works within a wide range of conditions
Specifications
Dimensions (L x W x H) 6 x 35 mm
For Use With (Equipment) VELP Magnetic stirrer, Heating magnetic stirrer and GDE
Width (Metric) 35 mm
Length (Metric) 6 mm