Learn More
Fisherbrand™ Stainless Steel Stirring Shafts with Propellers
Shop All Fisherbrand Products
Click to view available options
Size:
305mm
457mm
610mm
762mm
Includes:
Propellor, 108mm
Propellor, 63.5mm
Unit Size:
Each
Specifications
Specifications
| Includes | Propellor, 63.5mm |
| Quantity | 1 Each |
| Size | 610mm |
| Type | Stirring Shaft |