missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STK35 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-30644
This item is not returnable.
View return policy
Description
STK35 Polyclonal specifically detects STK35 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| STK35 | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Q8TDR2 | |
| STK35 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: TSLKRRHQNVVQFEECVLQRNGLAQRMSHGNKSSQLYLRLVET | |
| 0.1 mL | |
| Protein Kinase | |
| 140901 | |
| Human | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| bA550O8.2, CLIK-1, CLIK1Serine/threonine-protein kinase 35 L1, CLP-36 interacting kinase, CLP-36-interacting kinase 1, EC 2.7.11, EC 2.7.11.1, PDIK1, PDLIM1-interacting kinase 1, serine threonine kinase 35 long form, serine/threonine kinase 35, serine/threonine-protein kinase 35, STK35L1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction