Description
- Small, tapered plug used to seal the openings of test tubes, flasks and other laboratory glassware
- Preferable for applications that require tighter seal or greater degree of chemical resistance
- Prevent contamination of samples by protecting contents of laboratory glassware from air
- Allows experimenters to shake or mix solutions without spilling them
Specifications
Specifications
| Description | Two holed |
| Quantity | 5/Pk. |
| Color | Red |
| Material | Rubber |
| Product Type | Tapered Stopper (2 holes) |