Learn More
Invitrogen™ STX17 Monoclonal Antibody (K52003_13F1), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA556289
Description
Sequence of this protein is as follows: RRLEPAIQKF IKIVIPTDLE RLRKHQINIE KYQRCRIWDK LHEEHINAGR TVQQLRSNIR EIEKLCLKVR KDDLVLLKRM IDPVKEEASA ATAEFLQLHL ESVEELKKQF NDEETLLQPP LTRSMTVGGA FHTTEAEASS QSLTQIYALP EIPQDQNAAE SWETLEADLI ELSQLVTDFS LLVNSQQEKI DSIADHVNSA AVNVEEGTKN
SNAREs, soluble N-ethylmaleimide-sensitive factor-attachment protein receptors, are essential proteins for fusion of cellular membranes. SNAREs localized on opposing membranes assemble to form a trans-SNARE complex, an extended, parallel four alpha-helical bundle that drives membrane fusion. STX17 is a SNARE of the autophagosome involved in autophagy through the direct control of autophagosome membrane fusion with the lysosome membrane. STX17 may also play a role in the early secretory pathway where it may maintain the architecture of the endoplasmic reticulum-Golgi intermediate compartment/ERGIC and Golgi and/or regulate transport between the endoplasmic reticulum, the ERGIC and the Golgi. (Uniprot).
Specifications
| STX17 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| P56962 | |
| STX17 | |
| Recombinant human STX17 (immunogen range is 11-220). | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| K52003_13F1 | |
| Unconjugated | |
| STX17 | |
| 4833418L03Rik; 6330411F21Rik; 9030425C21Rik; AW048351; Stx17; syntaxin 17; syntaxin-17 | |
| Mouse | |
| Protein A | |
| RUO | |
| 55014 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.