missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ STX17 Monoclonal Antibody (K52003_13F1), Invitrogen™
Mouse Monoclonal Antibody
€ 493.00 - € 868.00
Specifications
| Antigen | STX17 |
|---|---|
| Clone | K52003_13F1 |
| Concentration | 1 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30196849
|
Invitrogen™
MA556289 |
200 μL |
€ 868.00
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30196368
|
Invitrogen™
MA555909 |
50 μL |
€ 493.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sequence of this protein is as follows: RRLEPAIQKF IKIVIPTDLE RLRKHQINIE KYQRCRIWDK LHEEHINAGR TVQQLRSNIR EIEKLCLKVR KDDLVLLKRM IDPVKEEASA ATAEFLQLHL ESVEELKKQF NDEETLLQPP LTRSMTVGGA FHTTEAEASS QSLTQIYALP EIPQDQNAAE SWETLEADLI ELSQLVTDFS LLVNSQQEKI DSIADHVNSA AVNVEEGTKN
STX17 Monoclonal antibody specifically detects STX17 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| STX17 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| 4833418L03Rik; 6330411F21Rik; 9030425C21Rik; AW048351; Stx17; syntaxin 17; syntaxin-17 | |
| Recombinant human STX17 (immunogen range is 11-220). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| STX17 |
| K52003_13F1 | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| P56962 | |
| 55014 | |
| IgG1 | |
| Protein A | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title