Learn More
SUCLA2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 12627-1-AP-20UL
 View more versions of this product
Product Code. 16850444
  • € 145.00 / 20µL
Description
SUCLA2 (succinate-CoA ligase, ADP-forming, beta subunit), also known as A-BETA, SCS-betaA or renal carcinoma antigen NY-REN-39, is a 463 amino acid mitochondrial matrix enzyme that belongs to the succinate/malate CoA ligase beta subunit family. Widely expressed, SUCLA2 dimerizes with the SCS alpha subunit to form SCS-A, an essential component of the tricarboxylic acid cycle. Defects in SUCLA2 may be involved in a group of autosomal recessive disorders known as Mitochondrial DNA depletion syndromes (MDSs) that are characterized by a decrease in mitochondrial DNA copy numbers in affected tissues.
Specifications
Specifications
SUCLA2
Polyclonal
Unconjugated
SUCLA2
A BETA, SCS betaA, SUCLA2
Rabbit
Antigen Affinity Chromatography
RUO
20916, 361071, 8803
-20°C
Liquid
Immunoprecipitation, Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot
0.25 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9P2R7, Q9Z2I9
Sucla2
SUCLA2 Fusion Protein Ag3319
20 μL
Primary
Rat, Mouse, Human