missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SUCLA2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
€ 145.00 - € 410.00
Specifications
| Antigen | SUCLA2 |
|---|---|
| Concentration | 0.25 mg/mL |
| Applications | Immunoprecipitation, Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16850444
|
Proteintech
12627-1-AP-20UL |
20 μL |
€ 145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16840444
|
Proteintech
12627-1-AP-150UL |
150 μL |
€ 410.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SUCLA2 (succinate-CoA ligase, ADP-forming, beta subunit), also known as A-BETA, SCS-betaA or renal carcinoma antigen NY-REN-39, is a 463 amino acid mitochondrial matrix enzyme that belongs to the succinate/malate CoA ligase beta subunit family. Widely expressed, SUCLA2 dimerizes with the SCS alpha subunit to form SCS-A, an essential component of the tricarboxylic acid cycle. Defects in SUCLA2 may be involved in a group of autosomal recessive disorders known as Mitochondrial DNA depletion syndromes (MDSs) that are characterized by a decrease in mitochondrial DNA copy numbers in affected tissues.Specifications
| SUCLA2 | |
| Immunoprecipitation, Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Rat, Mouse, Human | |
| Q9P2R7, Q9Z2I9 | |
| 20916, 361071, 8803 | |
| SUCLA2 Fusion Protein Ag3319 | |
| Primary | |
| -20°C | |
| SUCLA2 |
| 0.25 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| A BETA, SCS betaA, SUCLA2 | |
| Sucla2 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title