missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Swine Influenza A H1N1 NS1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41342
This item is not returnable.
View return policy
Description
Swine Influenza A H1N1 NS1 Polyclonal antibody specifically detects Swine Influenza A H1N1 NS1 in Virus samples. It is validated for ELISA.
Specifications
| Swine Influenza A H1N1 NS1 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| Antibody was raised against a synthetic peptide from the swine-Origin Influenza NS1. The immunogen is located within the last 50 amino acids of Swine H1N1 Nonstructural Protein 1. Amino Acid Squence: DENGRPSLPPE | |
| 0.1 mg | |
| Primary | |
| This antibody is specific for the Swine-Origin H1N1 influenza NS1 and will not recognize the corresponding NS1 sequence from the seasonal H1N1 influenza (A/South Australia/51/2005 (H1N1)). | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| ELISA | |
| 1 mg/mL | |
| ELISA | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 2828679 | |
| Virus | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction