missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Swine Influenza A H1N1 NS1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 168.00 - € 507.00
Specifications
| Antigen | Swine Influenza A H1N1 NS1 |
|---|---|
| Concentration | 1 mg/mL |
| Applications | ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18683419
|
Novus Biologicals
NBP2-41342-0.025mg |
0.025 mg |
€ 168.00
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18116918
|
Novus Biologicals
NBP2-41342 |
0.1 mg |
€ 507.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Swine Influenza A H1N1 NS1 Polyclonal antibody specifically detects Swine Influenza A H1N1 NS1 in Virus samples. It is validated for ELISA.Specifications
| Swine Influenza A H1N1 NS1 | |
| ELISA | |
| Unconjugated | |
| RUO | |
| PBS with 0.02% Sodium Azide | |
| Antibody was raised against a synthetic peptide from the swine-Origin Influenza NS1. The immunogen is located within the last 50 amino acids of Swine H1N1 Nonstructural Protein 1. Amino Acid Squence: DENGRPSLPPE | |
| Primary | |
| This antibody is specific for the Swine-Origin H1N1 influenza NS1 and will not recognize the corresponding NS1 sequence from the seasonal H1N1 influenza (A/South Australia/51/2005 (H1N1)). |
| 1 mg/mL | |
| Polyclonal | |
| Rabbit | |
| Virus | |
| 2828679 | |
| IgG | |
| Affinity Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title