Learn More
SYK Mouse, anti-Human, CoraLite Plus 488, Clone: 4C4A12, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech CL488-66721-20UL
 View more versions of this product
Product Code. 18018765
  • € 145.00 / 20µL
Description
The SYK antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human SYK. This antibody recognizes human antigen. The SYK antibody has been validated for the following applications: WB analysis. This antibody is coralite™ plus 488.
Specifications
Specifications
SYK
4C4A12
CoraLite Plus 488
Mouse
Primary
Human
155 kDa
IgG2b
Western Blot
1000 μg/mL
DKFZp313N1010, FLJ25043, FLJ37489, spleen tyrosine kinase, SYK, Tyrosine protein kinase SYK
20 μL
6850
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.