missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SYK Mouse, anti-Human, CoraLite Plus 488, Clone: 4C4A12, Proteintech
Mouse Monoclonal Antibody
€ 145.00 - € 450.00
Specifications
| Antigen | SYK |
|---|---|
| Clone | 4C4A12 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot |
| Conjugate | CoraLite Plus 488 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18010856
|
Proteintech
CL488-66721-100UL |
100 μL |
€ 450.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18018765
|
Proteintech
CL488-66721-20UL |
20 μL |
€ 145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The SYK antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human SYK. This antibody recognizes human antigen. The SYK antibody has been validated for the following applications: WB analysis. This antibody is coralite™ plus 488.
Specifications
| SYK | |
| 1000 μg/mL | |
| CoraLite Plus 488 | |
| Mouse | |
| 6850 | |
| IgG2b | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. |
| 4C4A12 | |
| Western Blot | |
| Liquid | |
| Human | |
| DKFZp313N1010, FLJ25043, FLJ37489, spleen tyrosine kinase, SYK, Tyrosine protein kinase SYK | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title