missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Synapsin II Recombinant Rabbit Monoclonal Antibody (23GB4660), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA552724
This item is not returnable.
View return policy
Description
Synapsins (synapsin I, II, III) are a family of synaptic vesicle phosphoproteins that have been implicated in modulation of neurotransmitter release and in synaptogenesis. Synapsin II (SYN2) is composed of two isoforms, synapsin IIa and IIb.
Specifications
| Synapsin II | |
| Recombinant Monoclonal | |
| Unconjugated | |
| SYN2 | |
| 2900074L19Rik; AI836018; AI841723; SYN2; synapsin 2; synapsin II; Synapsin-2; SYNII; SYNIIa; SYNIIb | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 6854 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB4660 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q92777 | |
| SYN2 | |
| A synthesized peptide derived from human Synapsin II (500-582AA). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction