Learn More
Syntaxin 16 Antibody (S09-9A8), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-19783-100ul
This item is not returnable.
View return policy
Description
Syntaxin 16 Monoclonal antibody specifically detects Syntaxin 16 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| Syntaxin 16 | |
| Monoclonal | |
| 0.3 mg/mL | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 8675 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| S09-9A8 | |
| Unconjugated | |
| MGC90328, Syn16, SYN16hsyn16, syntaxin 16, syntaxin-16 | |
| A synthetic peptide of human Syntaxin 16 (Uniprot # O14662) | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |