Learn More
Syntaxin 16 Antibody (S09-9A8), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-19783-100ul
 View more versions of this product
Product Code. 18655243
  • € 645.00 / 100µL
Description
Syntaxin 16 Monoclonal antibody specifically detects Syntaxin 16 in Human, Mouse, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Syntaxin 16
Monoclonal
0.3 mg/mL
50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA
Rabbit
Affinity purified
RUO
8675
Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.
IgG
Western Blot
S09-9A8
Unconjugated
MGC90328, Syn16, SYN16hsyn16, syntaxin 16, syntaxin-16
A synthetic peptide of human Syntaxin 16 (Uniprot # O14662)
100 μg
Primary
Human, Mouse, Rat