missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Syntaxin 1B Antibody (SR1555), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22030-100ul
This item is not returnable.
View return policy
Description
Syntaxin 1B Monoclonal antibody specifically detects Syntaxin 1B in Human, Mouse, Rat samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| Syntaxin 1B | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 112755 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
| Western Blot, Immunoprecipitation | |
| SR1555 | |
| Western Blot 1:1000-1:5000, Immunoprecipitation 1:50 | |
| FP17469, STX1B1, STX1B2, syntaxin 1B, syntaxin 1B1, syntaxin 1B2, syntaxin-1B, syntaxin-1B1, syntaxin-1B2 | |
| A synthesized peptide derived from human Syntaxin 1B (Uniprot #: P61266) | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction