missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Syntaxin 1B Antibody (SR1555), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 338.00 - € 449.00
Specifications
| Antigen | Syntaxin 1B |
|---|---|
| Clone | SR1555 |
| Dilution | Western Blot 1:1000-1:5000, Immunoprecipitation 1:50 |
| Applications | Western Blot, Immunoprecipitation |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18686114
|
Novus Biologicals
NBP3-22030-100ul |
100 μg |
€ 449.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18626504
|
Novus Biologicals
NBP3-22030-50ul |
50 μg |
€ 338.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Syntaxin 1B Monoclonal antibody specifically detects Syntaxin 1B in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
| Syntaxin 1B | |
| Western Blot 1:1000-1:5000, Immunoprecipitation 1:50 | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 112755 | |
| IgG | |
| Affinity purified |
| SR1555 | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| FP17469, STX1B1, STX1B2, syntaxin 1B, syntaxin 1B1, syntaxin 1B2, syntaxin-1B, syntaxin-1B1, syntaxin-1B2 | |
| A synthesized peptide derived from human Syntaxin 1B (Uniprot #: P61266) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title