missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SYPL1 Recombinant Rabbit Monoclonal Antibody (23GB4675), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA553787
This item is not returnable.
View return policy
Description
The synaptophysin-like protein 1, also known as pantophysin, is a homolog of the neuroendocrine-specific protein synaptophysin, with the highest level of homology across its four transmembrane domains. Unlike synaptophysin however, SYPL1 is ubiquitously expressed and found in small cytoplasmic transport vesicles regardless of their content. SYPL1 is thought to play a multifunctional role in vesicle biogenesis and transport and is a component of adipocyte transport vesicles, and thus may be involved in adipocyte secretion. SYPL1 also interacts with vesicle-associated membrane protein 2 (VAMP-2) in adipocytes and associates with GLUT4-containing vesicles.
Specifications
| SYPL1 | |
| Recombinant Monoclonal | |
| 0.50 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q16563 | |
| SYPL1 | |
| A synthesized peptide derived from human SYPL1 (150-259AA). | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB4675 | |
| Unconjugated | |
| SYPL1 | |
| AI314763; AI604763; D12Ertd446e; H-SP1; Pan I; PanI; Pantophysin; Pphn; synaptophysin like 1; synaptophysin-like 1; synaptophysin-like protein; synaptophysin-like protein 1; SYPL; Sypl1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 6856 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction