missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SYT11 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 221.00 - € 305.00
Specifications
| Antigen | SYT11 |
|---|---|
| Dilution | Western Blot 1:500-1:2000, Immunoprecipitation 1-5 μL/mg of lysate, Immunohistochemistry-Paraffin 1:50-1:200 |
| Applications | Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
SYT11 Polyclonal antibody specifically detects SYT11 in Human samples. It is validated for Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)Specifications
| SYT11 | |
| Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Neuronal Cell Markers, Neurotransmission | |
| PBS (pH 7.0) | |
| 23208 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Western Blot 1:500-1:2000, Immunoprecipitation 1-5 μL/mg of lysate, Immunohistochemistry-Paraffin 1:50-1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| DKFZp781D015, KIAA0080synaptotagmin 12, MGC10881, MGC17226, synaptotagmin XISYT12, synaptotagmin-11, SytXI | |
| Produced in rabbits immunized with E. coli-derived Human SYT11 fragment. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title