Learn More
TAF15 Rabbit, anti-Human, Clone: 3L13, Proteintech
Rabbit Recombinant Monoclonal Antibody
Brand:  Proteintech 82860-1-RR-20UL
 View more versions of this product
Product Code. 18027795
  • € 149.00 / 20µL
Description
The TAF15 antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human TAF15. This antibody recognizes human, mouse, rat antigen. The TAF15 antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
Specifications
TAF15
3L13
hTAFII68, RBP56, RNA binding protein 56, TAF(II)68, TAF15, TAF2N, TAFII68
20 μL
8148
Store at -20°C. Stable for one year after shipment.
Liquid
Western Blot, Immunofluorescence, ELISA
800 μg/mL
Rabbit
Primary
Human, Mouse, Rat
150 kDa