Learn More
TAF15 Rabbit, anti-Human, Clone: 3L13, Proteintech
Rabbit Recombinant Monoclonal Antibody
Brand: Proteintech 82860-1-RR-20UL
Description
The TAF15 antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human TAF15. This antibody recognizes human, mouse, rat antigen. The TAF15 antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
| TAF15 | |
| 3L13 | |
| hTAFII68, RBP56, RNA binding protein 56, TAF(II)68, TAF15, TAF2N, TAFII68 | |
| 20 μL | |
| 8148 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
| Western Blot, Immunofluorescence, ELISA | |
| 800 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse, Rat | |
| 150 kDa | |
| IgG |