missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAF15 Rabbit, anti-Human, Clone: 3L13, Proteintech
Rabbit Recombinant Monoclonal Antibody
€ 149.00 - € 439.00
Specifications
| Antigen | TAF15 |
|---|---|
| Clone | 3L13 |
| Concentration | 800 μg/mL |
| Applications | Western Blot, Immunofluorescence, ELISA |
| Form | Liquid |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18027795
|
Proteintech
82860-1-RR-20UL |
20 μL |
€ 149.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18016385
|
Proteintech
82860-1-RR-100UL |
100 μL |
€ 439.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The TAF15 antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human TAF15. This antibody recognizes human, mouse, rat antigen. The TAF15 antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
| TAF15 | |
| 800 μg/mL | |
| Liquid | |
| Human, Mouse, Rat | |
| hTAFII68, RBP56, RNA binding protein 56, TAF(II)68, TAF15, TAF2N, TAFII68 | |
| Primary |
| 3L13 | |
| Western Blot, Immunofluorescence, ELISA | |
| Rabbit | |
| 8148 | |
| IgG | |
| Store at -20°C. Stable for one year after shipment. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title